Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTCCCTCAATGGATTGGTTTACA[C/T]AGGTCAAGTAAGTAAATAGAGATTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
NHLRC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NHLRC3 - NHL repeat containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012754.3 | 559 | Missense Mutation | ACA,ATA | T,I 77 | NP_001012772.1 | |
NM_001017370.2 | 559 | Missense Mutation | ACA,ATA | T,I 77 | NP_001017370.1 |
PROSER1 - proline and serine rich 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |