Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCAACACCCAGGGCAGGTGGTA[C/T]CCATTGTAAAGGAAGTAAAACACAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604418 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GJB6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GJB6 - gap junction protein beta 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001110219.2 | 793 | Silent Mutation | GGA,GGG | G,G 160 | NP_001103689.1 | |
NM_001110220.2 | 793 | Silent Mutation | GGA,GGG | G,G 160 | NP_001103690.1 | |
NM_001110221.2 | 793 | Silent Mutation | GGA,GGG | G,G 160 | NP_001103691.1 | |
NM_006783.4 | 793 | Silent Mutation | GGA,GGG | G,G 160 | NP_006774.2 | |
XM_017020357.1 | 793 | Silent Mutation | GGA,GGG | G,G 160 | XP_016875846.1 | |
XM_017020358.1 | 793 | Silent Mutation | GGA,GGG | G,G 160 | XP_016875847.1 | |
XM_017020359.1 | 793 | Silent Mutation | GGA,GGG | G,G 160 | XP_016875848.1 | |
XM_017020360.1 | 793 | Silent Mutation | GGA,GGG | G,G 160 | XP_016875849.1 |