Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTAGTCCTGGTGGATCTGCAGTA[C/T]GAAGGTGAGTCCTGCCCTGCCCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601184 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJC3 - DnaJ heat shock protein family (Hsp40) member C3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006260.4 | 195 | Silent Mutation | TAC,TAT | Y,Y 26 | NP_006251.1 | |
XM_011521104.2 | 195 | Silent Mutation | TAC,TAT | Y,Y 26 | XP_011519406.1 | |
XM_011521105.1 | 195 | Intron | XP_011519407.1 | |||
XM_017020674.1 | 195 | Intron | XP_016876163.1 | |||
XM_017020675.1 | 195 | Intron | XP_016876164.1 |
DNAJC3-AS1 - DNAJC3 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |