Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTCATGGGCTGTGATGATGGAA[A/T]GGCTTGTAAAATAAACTTCTGAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 189973 | ||||||||||||||||||||
Literature Links: |
ELF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ELF1 - E74 like ETS transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145353.1 | 2013 | Missense Mutation | ATT,TTT | I,F 447 | NP_001138825.1 | |
NM_172373.3 | 2013 | Missense Mutation | ATT,TTT | I,F 471 | NP_758961.1 | |
XM_005266276.1 | 2013 | Missense Mutation | ATT,TTT | I,F 471 | XP_005266333.1 | |
XM_005266277.3 | 2013 | Missense Mutation | ATT,TTT | I,F 471 | XP_005266334.1 | |
XM_011534950.1 | 2013 | Missense Mutation | ATT,TTT | I,F 471 | XP_011533252.1 | |
XM_011534951.2 | 2013 | Missense Mutation | ATT,TTT | I,F 412 | XP_011533253.1 | |
XM_017020409.1 | 2013 | Missense Mutation | ATT,TTT | I,F 471 | XP_016875898.1 | |
XM_017020410.1 | 2013 | Missense Mutation | ATT,TTT | I,F 471 | XP_016875899.1 | |
XM_017020411.1 | 2013 | Missense Mutation | ATT,TTT | I,F 471 | XP_016875900.1 | |
XM_017020412.1 | 2013 | Missense Mutation | ATT,TTT | I,F 471 | XP_016875901.1 | |
XM_017020413.1 | 2013 | Missense Mutation | ATT,TTT | I,F 471 | XP_016875902.1 |