Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTTTTTTTTCAAGAATCCTCTA[C/T]GCCCCCTACAAGTTCATTCGTTCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603803 | ||||||||||||||||||||
Literature Links: |
DACH1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DACH1 - dachshund family transcription factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004392.6 | 2044 | UTR 3 | NP_004383.4 | |||
NM_080759.5 | 2044 | UTR 3 | NP_542937.3 | |||
NM_080760.5 | 2044 | UTR 3 | NP_542938.3 | |||
XM_011534939.2 | 2044 | UTR 3 | XP_011533241.1 | |||
XM_011534940.2 | 2044 | Intron | XP_011533242.1 | |||
XM_011534941.2 | 2044 | Intron | XP_011533243.1 | |||
XM_011534942.2 | 2044 | UTR 3 | XP_011533244.1 | |||
XM_017020396.1 | 2044 | UTR 3 | XP_016875885.1 |