Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATACAAACTTAAAGGACTCTCTCT[C/G]AGGTTCTTTGACTGACGTCCACTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602148 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC00403 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC00403 - long intergenic non-protein coding RNA 403 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SOX1 - SRY-box 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005986.2 | 3526 | UTR 3 | NP_005977.2 |