Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAGCCAAAAGCTGTAAGCGTTTGC[A/G]TAGTAATTGCAGGTCCCACGGCCGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 120130 | ||||||||||||||||||||
Literature Links: |
COL4A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
COL4A1 - collagen type IV alpha 1 chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303110.1 | 12937 | Intron | NP_001290039.1 | |||
NM_001845.5 | 12937 | Silent Mutation | TAC,TAT | Y,Y 1625 | NP_001836.3 | |
XM_011521048.2 | 12937 | Silent Mutation | TAC,TAT | Y,Y 1561 | XP_011519350.1 |