Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGTAAAGTGCAGAGAAATCTGGA[A/T]CGAATGTCTTTGGCTACATTCCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 605741 | ||||||||||||||||||||
Literature Links: |
GPR183 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR183 - G protein-coupled receptor 183 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004951.4 | 1030 | Missense Mutation | ATC,TTC | I,F 286 | NP_004942.1 | |
XM_017020405.1 | 1030 | Missense Mutation | ATC,TTC | I,F 286 | XP_016875894.1 |
UBAC2 - UBA domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144072.1 | 1030 | Intron | NP_001137544.1 | |||
NM_177967.3 | 1030 | Intron | NP_808882.1 | |||
XM_006719948.3 | 1030 | Intron | XP_006720011.1 | |||
XM_011521082.2 | 1030 | Intron | XP_011519384.1 | |||
XM_011521083.2 | 1030 | Intron | XP_011519385.1 | |||
XM_011521084.2 | 1030 | Intron | XP_011519386.1 | |||
XM_017020553.1 | 1030 | Intron | XP_016876042.1 |