Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAGCCATTGCAGCTGCTCACGGAG[A/G]AGTAGCCCCCCAGGGCACTCATCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 136533 | ||||||||||||||||||||
Literature Links: |
FOXO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FOXO1 - forkhead box O1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002015.3 | 2121 | Missense Mutation | TCC,TTC | S,F 579 | NP_002006.2 | |
XM_011535008.2 | 2121 | Missense Mutation | TCC,TTC | S,F 398 | XP_011533310.1 | |
XM_011535010.2 | 2121 | Missense Mutation | TCC,TTC | S,F 342 | XP_011533312.1 |