Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCTGGGAGGAGCAGCTGAACCGA[A/G]ACTAGCTACCTGACGATATGAGGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607519 | ||||||||||||||||||||
Literature Links: |
PARP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PARP4 - poly(ADP-ribose) polymerase family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006437.3 | 4371 | Missense Mutation | CTC,TTC | L,F 1350 | NP_006428.2 | |
XM_011534931.1 | 4371 | Missense Mutation | CTC,TTC | L,F 1397 | XP_011533233.1 | |
XM_011534932.2 | 4371 | Missense Mutation | CTC,TTC | L,F 1350 | XP_011533234.1 |