Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAAATTTGATGTGTCTATCTTC[A/G]TGAGCAGTTATTGTAACAGGAAGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614766 | ||||||||||||||||||||
Literature Links: |
STRN3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
STRN3 - striatin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083893.1 | 2316 | Silent Mutation | CAC,CAT | H,H 700 | NP_001077362.1 | |
NM_014574.3 | 2316 | Silent Mutation | CAC,CAT | H,H 616 | NP_055389.3 | |
XM_005267569.3 | 2316 | Silent Mutation | CAC,CAT | H,H 663 | XP_005267626.1 | |
XM_005267570.3 | 2316 | Silent Mutation | CAC,CAT | H,H 653 | XP_005267627.1 |