Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTAATCCAGTGGCAGAGAATTTCC[A/G]AATGGAAGAAGTCTATTTACCAGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608642 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTGR2 - prostaglandin reductase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146154.1 | 279 | Missense Mutation | CAA,CGA | Q,R 24 | NP_001139626.1 | |
NM_001146155.1 | 279 | Missense Mutation | CAA,CGA | Q,R 24 | NP_001139627.1 | |
NM_152444.2 | 279 | Missense Mutation | CAA,CGA | Q,R 24 | NP_689657.1 |