Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAGGTCTAGTAAGACCCAAAGAG[A/G]AAATTTGTTTTGCACCTTTTCTGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608467 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STON2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STON2 - stonin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256430.1 | 5675 | Intron | NP_001243359.1 | |||
NM_033104.3 | 5675 | UTR 3 | NP_149095.2 | |||
XM_006720279.1 | 5675 | UTR 3 | XP_006720342.1 | |||
XM_011537240.1 | 5675 | Intron | XP_011535542.1 | |||
XM_011537241.1 | 5675 | Intron | XP_011535543.1 | |||
XM_011537242.1 | 5675 | UTR 3 | XP_011535544.1 |