Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCTCCAAGAAAGTCTTGCCATGAT[C/T]GGATAGGACCCCCAGATAAATATTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616003 MIM: 603885 | ||||||||||||||||||||
Literature Links: |
APOPT1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
APOPT1 - apoptogenic 1, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302652.1 | 208 | Missense Mutation | CGG,TGG | R,W 68 | NP_001289581.1 | |
NM_001302653.1 | 208 | Missense Mutation | CGG,TGG | R,W 68 | NP_001289582.1 | |
NM_001302654.1 | 208 | Missense Mutation | CGG,TGG | R,W 68 | NP_001289583.1 | |
NM_032374.4 | 208 | Missense Mutation | CGG,TGG | R,W 68 | NP_115750.2 |
BAG5 - BCL2 associated athanogene 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |