Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAAAACTTTATGTACAGAATGCA[A/G]GAAAAAAACATAAGAGGCACTTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603812 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DCAF5 - DDB1 and CUL4 associated factor 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001284206.1 | 5697 | UTR 3 | NP_001271135.1 | |||
NM_001284207.1 | 5697 | UTR 3 | NP_001271136.1 | |||
NM_001284208.1 | 5697 | Intron | NP_001271137.1 | |||
NM_003861.2 | 5697 | UTR 3 | NP_003852.1 | |||
XM_006720297.2 | 5697 | UTR 3 | XP_006720360.1 | |||
XM_006720298.2 | 5697 | UTR 3 | XP_006720361.1 | |||
XM_006720299.3 | 5697 | UTR 3 | XP_006720362.1 | |||
XM_006720300.2 | 5697 | UTR 3 | XP_006720363.1 | |||
XM_011537278.1 | 5697 | UTR 3 | XP_011535580.1 | |||
XM_011537279.2 | 5697 | UTR 3 | XP_011535581.1 | |||
XM_011537280.2 | 5697 | UTR 3 | XP_011535582.1 | |||
XM_017021733.1 | 5697 | UTR 3 | XP_016877222.1 | |||
XM_017021734.1 | 5697 | UTR 3 | XP_016877223.1 | |||
XM_017021735.1 | 5697 | UTR 3 | XP_016877224.1 | |||
XM_017021736.1 | 5697 | UTR 3 | XP_016877225.1 | |||
XM_017021737.1 | 5697 | UTR 3 | XP_016877226.1 | |||
XM_017021738.1 | 5697 | UTR 3 | XP_016877227.1 | |||
XM_017021739.1 | 5697 | UTR 3 | XP_016877228.1 |