Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAAGGAGTCTCCAGTTACTGAGTG[A/G]CCACTCCGTGCCAGCTCAGCCCACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612498 | ||||||||||||||||||||
Literature Links: |
ADSSL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADSSL1 - adenylosuccinate synthase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320424.1 | 161 | UTR 5 | NP_001307353.1 | |||
NM_152328.4 | 161 | Intron | NP_689541.1 | |||
NM_199165.2 | 161 | Missense Mutation | GAC,GGC | D,G 49 | NP_954634.1 | |
XM_006720026.3 | 161 | Intron | XP_006720089.1 | |||
XM_011536412.1 | 161 | Missense Mutation | GAC,GGC | D,G 49 | XP_011534714.1 | |
XM_011536414.2 | 161 | Intron | XP_011534716.1 |
LOC388022 - uncharacterized LOC388022 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |