Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACTCAGTCTCGCTTTCGTCCTTG[C/T]GCCTCAGGATGGTGAGGGCGTCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606455 MIM: 613504 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R13B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R13B - protein phosphatase 1 regulatory subunit 13B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015316.2 | 3944 | Missense Mutation | CAC,CGC | H,R 1051 | NP_056131.2 | |
XM_005267487.4 | 3944 | Missense Mutation | CAC,CGC | H,R 1119 | XP_005267544.1 | |
XM_011536593.2 | 3944 | Missense Mutation | CAC,CGC | H,R 1154 | XP_011534895.1 | |
XM_017021116.1 | 3944 | Missense Mutation | CAC,CGC | H,R 1086 | XP_016876605.1 | |
XM_017021117.1 | 3944 | Missense Mutation | CAC,CGC | H,R 1083 | XP_016876606.1 | |
XM_017021118.1 | 3944 | Missense Mutation | CAC,CGC | H,R 1083 | XP_016876607.1 | |
XM_017021119.1 | 3944 | Missense Mutation | CAC,CGC | H,R 834 | XP_016876608.1 |
ZFYVE21 - zinc finger FYVE-type containing 21 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |