Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAAATTTATGAACTTGATCTTC[A/G]GCCGGTTGATGCCAAATCGCCTTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602680 | ||||||||||||||||||||
Literature Links: |
ARHGAP5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGAP5 - Rho GTPase activating protein 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030055.1 | 2188 | Missense Mutation | CAG,CGG | Q,R 635 | NP_001025226.1 | |
NM_001173.2 | 2188 | Missense Mutation | CAG,CGG | Q,R 635 | NP_001164.2 | |
XM_005267635.3 | 2188 | Missense Mutation | CAG,CGG | Q,R 635 | XP_005267692.1 | |
XM_005267636.3 | 2188 | Missense Mutation | CAG,CGG | Q,R 635 | XP_005267693.1 | |
XM_017021287.1 | 2188 | Missense Mutation | CAG,CGG | Q,R 635 | XP_016876776.1 | |
XM_017021288.1 | 2188 | Missense Mutation | CAG,CGG | Q,R 635 | XP_016876777.1 |