Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGTCTTTTCATGATATAGGCCA[C/T]CAATCAAATAGTCCAAAGCATCTTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
WDR89 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
WDR89 - WD repeat domain 89 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008726.2 | 1598 | Missense Mutation | GAT,GGT | D,G 279 | NP_001008726.1 | |
NM_001258272.1 | 1598 | Missense Mutation | GAT,GGT | D,G 279 | NP_001245201.1 | |
NM_080666.3 | 1598 | Missense Mutation | GAT,GGT | D,G 279 | NP_542397.1 | |
XM_011536383.2 | 1598 | Missense Mutation | GAT,GGT | D,G 317 | XP_011534685.1 | |
XM_011536385.2 | 1598 | Missense Mutation | GAT,GGT | D,G 279 | XP_011534687.1 |