Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGACTATCTTCTCATGGAGGAAGA[A/G]TTCATTAGAAATCAGGAACAAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602706 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMC1 - proteasome 26S subunit, ATPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002802.2 | 1798 | Intron | NP_002793.2 | |||
XM_017021470.1 | 1798 | Silent Mutation | GAA,GAG | E,E 4 | XP_016876959.1 | |
XM_017021471.1 | 1798 | Intron | XP_016876960.1 |