Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACATCTACCACCCACAGTGTCTG[C/T]GGTGAGCAGGGGCAGATGTGAGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614716 MIM: 615195 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CARMIL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CARMIL3 - capping protein regulator and myosin 1 linker 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_138360.3 | 176 | Silent Mutation | TGC,TGT | C,C 153 | NP_612369.3 | |
XM_011537300.2 | 176 | Silent Mutation | TGC,TGT | C,C 56 | XP_011535602.1 | |
XM_017021751.1 | 176 | Silent Mutation | TGC,TGT | C,C 33 | XP_016877240.1 |
DHRS4L1 - dehydrogenase/reductase 4 like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |