Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTATGAAGAAGGTTCCAACAAAAG[A/G]CTTTTTAATGTTGATCGGCATGTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176843 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PSMA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PSMA3 - proteasome subunit alpha 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002788.3 | 314 | Silent Mutation | AGA,AGG | R,R 66 | NP_002779.1 | |
NM_152132.2 | 314 | Silent Mutation | AGA,AGG | R,R 66 | NP_687033.1 |