Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGTGGTGGCGACGACGATGGACC[A/G]GGAGCCAGTGGGCGGTGTGGAACGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM177A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM177A1 - family with sequence similarity 177 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079519.1 | 467 | Missense Mutation | CAG,CGG | Q,R 3 | NP_001072987.1 | |
NM_001289022.1 | 467 | Missense Mutation | CAG,CGG | Q,R 3 | NP_001275951.1 | |
NM_173607.3 | 467 | Missense Mutation | CAG,CGG | Q,R 26 | NP_775878.2 |