Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAGGTAGTGTACACTCCAGTTTT[C/T]TGGTTCCTTTTAAAACAAAGGGGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610858 MIM: 605399 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C14orf166 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C14orf166 - chromosome 14 open reading frame 166 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NID2 - nidogen 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007361.3 | 4121 | Missense Mutation | AAA,AGA | K,R 1296 | NP_031387.3 | |
XM_005267405.4 | 4121 | Missense Mutation | AAA,AGA | K,R 1323 | XP_005267462.1 | |
XM_005267406.4 | 4121 | Missense Mutation | AAA,AGA | K,R 1275 | XP_005267463.1 | |
XM_005267407.4 | 4121 | Missense Mutation | AAA,AGA | K,R 1248 | XP_005267464.1 |