Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTCGCCGAGCTCAACAGGAGTCAG[C/T]GCAGGCCGAGAATCACCTCAATGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614068 MIM: 190230 | ||||||||||||||||||||
Literature Links: |
IFT43 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IFT43 - intraflagellar transport 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001102564.1 | 126 | Missense Mutation | GCG,GTG | A,V 31 | NP_001096034.1 | |
NM_001255995.1 | 126 | Missense Mutation | GCG,GTG | A,V 31 | NP_001242924.1 | |
NM_052873.2 | 126 | Missense Mutation | GCG,GTG | A,V 31 | NP_443105.2 |
TGFB3 - transforming growth factor beta 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |