Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAGAAGGAAAGAAAGTGGTTAC[A/C]GGGCAACGGGTCAGTGAGATCTCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603196 MIM: 614766 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COCH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COCH - cochlin | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100506071 - uncharacterized LOC100506071 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STRN3 - striatin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001083893.1 | 3307 | UTR 3 | NP_001077362.1 | |||
NM_014574.3 | 3307 | UTR 3 | NP_055389.3 | |||
XM_005267569.3 | 3307 | UTR 3 | XP_005267626.1 | |||
XM_005267570.3 | 3307 | UTR 3 | XP_005267627.1 |