Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGGAATTCTCTGTGTATGGGAAC[A/G]TAGTATATGCTTCTGTATCGAGCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603196 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COCH PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COCH - cochlin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135058.1 | 646 | Missense Mutation | ATA,GTA | I,V 65 | NP_001128530.1 | |
NM_004086.2 | 646 | Missense Mutation | ATA,GTA | I,V 65 | NP_004077.1 | |
XM_017021071.1 | 646 | Missense Mutation | ATA,GTA | I,V 130 | XP_016876560.1 | |
XM_017021072.1 | 646 | Missense Mutation | ATA,GTA | I,V 130 | XP_016876561.1 | |
XM_017021073.1 | 646 | Intron | XP_016876562.1 |
LOC100506071 - uncharacterized LOC100506071 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |