Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTTTCAGGGCTCAGTAAAGAGAC[A/C]AGCACTATATGCCTGTAGTACCTGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 613816 | ||||||||||||||||||||
Literature Links: |
GON7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GON7 - GON7, KEOPS complex subunit homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBR7 - ubiquitin protein ligase E3 component n-recognin 7 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_175748.3 | 403 | Missense Mutation | CAA,CCA | Q,P 56 | NP_786924.2 |