Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCCAGAAGAAACTTCACTACTGA[A/C]ATATTCCAAGAGGACTTGATGCTGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609066 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AJUBA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AJUBA - ajuba LIM protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C14orf93 - chromosome 14 open reading frame 93 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130706.2 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | NP_001124178.1 | |
NM_001130708.2 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | NP_001124180.1 | |
NM_001282968.1 | 1181 | Missense Mutation | ATG,ATT | M,I 154 | NP_001269897.1 | |
NM_001282969.1 | 1181 | Missense Mutation | ATG,ATT | M,I 154 | NP_001269898.1 | |
NM_001282970.1 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | NP_001269899.1 | |
NM_021944.3 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | NP_068763.2 | |
XM_005267971.1 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | XP_005268028.1 | |
XM_005267972.1 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | XP_005268029.1 | |
XM_006720231.1 | 1181 | Missense Mutation | ATG,ATT | M,I 154 | XP_006720294.1 | |
XM_006720232.2 | 1181 | Missense Mutation | ATG,ATT | M,I 154 | XP_006720295.1 | |
XM_011537059.1 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | XP_011535361.1 | |
XM_011537060.2 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | XP_011535362.1 | |
XM_011537062.1 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | XP_011535364.1 | |
XM_017021569.1 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | XP_016877058.1 | |
XM_017021570.1 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | XP_016877059.1 | |
XM_017021571.1 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | XP_016877060.1 | |
XM_017021572.1 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | XP_016877061.1 | |
XM_017021573.1 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | XP_016877062.1 | |
XM_017021574.1 | 1181 | Missense Mutation | ATG,ATT | M,I 334 | XP_016877063.1 | |
XM_017021575.1 | 1181 | Missense Mutation | ATG,ATT | M,I 154 | XP_016877064.1 | |
XM_017021576.1 | 1181 | Missense Mutation | ATG,ATT | M,I 154 | XP_016877065.1 | |
XM_017021577.1 | 1181 | Missense Mutation | ATG,ATT | M,I 154 | XP_016877066.1 |