Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTGGCATCACCTTATCCATTGCA[C/T]TTGCCTTTTAAACCACGTCGTCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615260 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01599 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01599 - long intergenic non-protein coding RNA 1599 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VCPKMT - valosin containing protein lysine methyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040662.1 | 856 | Intron | NP_001035752.1 | |||
NM_024558.2 | 856 | Intron | NP_078834.2 | |||
XM_005268060.3 | 856 | Intron | XP_005268117.1 | |||
XM_017021640.1 | 856 | UTR 3 | XP_016877129.1 | |||
XM_017021641.1 | 856 | UTR 3 | XP_016877130.1 | |||
XM_017021642.1 | 856 | Intron | XP_016877131.1 |