Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACTGGAGTCTGAGGATCGGTATC[A/G]TCCATCAGGGTAGAGCTGCCACATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186960 | ||||||||||||||||||||
Literature Links: |
TCL1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TCL1A - T-cell leukemia/lymphoma 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098725.1 | 377 | Nonsense Mutation | CGA,TGA | R,* 83 | NP_001092195.1 | |
NM_021966.2 | 377 | Nonsense Mutation | CGA,TGA | R,* 83 | NP_068801.1 | |
XM_017021676.1 | 377 | Nonsense Mutation | CGA,TGA | R,* 83 | XP_016877165.1 | |
XM_017021677.1 | 377 | Nonsense Mutation | CGA,TGA | R,* 83 | XP_016877166.1 |