Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAGTGGTAAGAAAGGTAATAAAG[C/T]ATTTAGTTGTGCCTTTAAGTAGGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616510 MIM: 615814 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNPNAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNPNAT1 - glucosamine-phosphate N-acetyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198066.3 | 2626 | UTR 3 | NP_932332.1 | |||
XM_005268012.2 | 2626 | UTR 3 | XP_005268069.1 | |||
XM_006720238.3 | 2626 | UTR 3 | XP_006720301.1 |
STYX - serine/threonine/tyrosine interacting protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |