Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCTCCTCACGGATCTGCACCAG[C/T]AACTGAAAGAGGTTTGGGGCTGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610062 MIM: 604010 | ||||||||||||||||||||
Literature Links: |
DNAL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNAL1 - dynein axonemal light chain 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ELMSAN1 - ELM2 and Myb/SANT domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PNMA1 - paraneoplastic Ma antigen 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006029.4 | 1758 | Silent Mutation | TTA,TTG | L,L 324 | NP_006020.4 |