Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTCAGCCCAATCTGGAACAAAATC[A/G]TAGGCCTCAGCCACAATAGAAGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603910 MIM: 610844 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF3J PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF3J - eukaryotic translation initiation factor 3 subunit J | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPG11 - spastic paraplegia 11 (autosomal recessive) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160227.1 | 6910 | Silent Mutation | NP_001153699.1 | |||
NM_025137.3 | 6910 | Silent Mutation | NP_079413.3 | |||
XM_006720700.1 | 6910 | Silent Mutation | XP_006720763.1 | |||
XM_006720701.3 | 6910 | Intron | XP_006720764.1 | |||
XM_017022634.1 | 6910 | Silent Mutation | XP_016878123.1 | |||
XM_017022635.1 | 6910 | Intron | XP_016878124.1 | |||
XM_017022636.1 | 6910 | Silent Mutation | XP_016878125.1 |