Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGACCCTTTCATCCAGGTTTAAGC[C/T]TACTGCCTCGTCCATTCCCACAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605715 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD276 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD276 - CD276 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024736.1 | Intron | NP_001019907.1 | ||||
NM_025240.2 | Intron | NP_079516.1 | ||||
XM_005254700.4 | Intron | XP_005254757.1 | ||||
XM_011522095.2 | Intron | XP_011520397.1 | ||||
XM_011522096.2 | Intron | XP_011520398.1 | ||||
XM_017022638.1 | Intron | XP_016878127.1 |