Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGAAGGATTGCTTTCTCATTATTTG[C/T]CAGATCTGCAGAGATCAAAACAAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604310 | ||||||||||||||||||||
Literature Links: |
BLOC1S6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BLOC1S6 - biogenesis of lysosomal organelles complex 1 subunit 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001311255.1 | 339 | Missense Mutation | CCA,TCA | P,S 67 | NP_001298184.1 | |
NM_001311256.1 | 339 | Missense Mutation | CCA,TCA | P,S 67 | NP_001298185.1 | |
NM_012388.3 | 339 | Missense Mutation | CCA,TCA | P,S 62 | NP_036520.1 |