Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAATGAAGTGGCGTGTAAGAAGAC[G/T]AAAATAAAGAAAACTTGCAGAAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600215 | ||||||||||||||||||||
Literature Links: |
MFAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MFAP1 - microfibrillar associated protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR76 - WD repeat domain 76 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001167941.1 | 261 | Silent Mutation | ACG,ACT | T,T 20 | NP_001161413.1 | |
NM_024908.3 | 261 | Silent Mutation | ACG,ACT | T,T 84 | NP_079184.2 | |
XM_005254680.3 | 261 | Silent Mutation | ACG,ACT | T,T 20 | XP_005254737.1 |