Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGCCCGGGGAACATGAGGTGGCG[C/T]GGAGTCTCGCCTTACTGGCACGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602904 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAPK6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MAPK6 - mitogen-activated protein kinase 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002748.3 | Intron | NP_002739.1 | ||||
XM_005254537.2 | Intron | XP_005254594.1 | ||||
XM_005254538.2 | Intron | XP_005254595.1 | ||||
XM_005254539.3 | Intron | XP_005254596.1 | ||||
XM_011521782.1 | Intron | XP_011520084.1 |