Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTTTTTCACAGCAGACACAGCCCG[C/T]GCAGGGGCTGCTGAAAGGCCACCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602416 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AP3S2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AP3S2 - adaptor related protein complex 3 sigma 2 subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005829.4 | 913 | Silent Mutation | GCA,GCG | A,A 159 | NP_005820.1 |
C15orf38-AP3S2 - C15orf38-AP3S2 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199058.1 | 913 | Silent Mutation | GCA,GCG | A,A 360 | NP_001185987.1 |