Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCATTGTTTTACAAGCATTGATC[A/G]TTTCATCGGGGAAATAGCGTGTAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603576 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRPM1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRPM1 - transient receptor potential cation channel subfamily M member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252020.1 | 4714 | Missense Mutation | ACG,ATG | T,M 1467 | NP_001238949.1 | |
NM_001252024.1 | 4714 | Missense Mutation | ACG,ATG | T,M 1450 | NP_001238953.1 | |
NM_001252030.1 | 4714 | Intron | NP_001238959.1 | |||
NM_002420.5 | 4714 | Missense Mutation | ACG,ATG | T,M 1428 | NP_002411.3 |