Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAACTGTGTCATGTAGGCTTGTAA[C/T]ATACACAGACACAGGAGTTTCCACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 167405 MIM: 603521 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC100507472 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC100507472 - uncharacterized LOC100507472 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCSK6 - proprotein convertase subtilisin/kexin type 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291309.1 | 4147 | UTR 3 | NP_001278238.1 | |||
NM_002570.4 | 4147 | UTR 3 | NP_002561.1 | |||
NM_138319.3 | 4147 | UTR 3 | NP_612192.1 | |||
NM_138322.3 | 4147 | Intron | NP_612195.1 | |||
NM_138323.2 | 4147 | Intron | NP_612196.1 | |||
NM_138324.2 | 4147 | Intron | NP_612197.1 | |||
NM_138325.3 | 4147 | Intron | NP_612198.2 |
SNRPA1 - small nuclear ribonucleoprotein polypeptide A' | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |