Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTCTACTGGTCCTTAGACAGAG[A/G]TAGAGACAAGTCAGGGGTTCCAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
GRAMD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GRAMD2 - GRAM domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001012642.2 | Intron | NP_001012660.1 | ||||
XM_011521327.2 | Intron | XP_011519629.1 | ||||
XM_011521328.2 | Intron | XP_011519630.1 | ||||
XM_011521329.2 | Intron | XP_011519631.1 | ||||
XM_011521330.2 | Intron | XP_011519632.1 | ||||
XM_011521331.2 | Intron | XP_011519633.1 | ||||
XM_011521332.2 | Intron | XP_011519634.1 | ||||
XM_011521333.2 | Intron | XP_011519635.1 | ||||
XM_017021997.1 | Intron | XP_016877486.1 | ||||
XM_017021998.1 | Intron | XP_016877487.1 |