Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATTCACTTGTTCCTGAAAATACT[A/G]AAGGTAAAGGTGATCTGTAGATTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612820 MIM: 615176 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NPTN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NPTN - neuroplastin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161363.1 | Intron | NP_001154835.1 | ||||
NM_001161364.1 | Intron | NP_001154836.1 | ||||
NM_012428.3 | Intron | NP_036560.1 | ||||
NM_017455.3 | Intron | NP_059429.1 | ||||
XM_011521452.2 | Intron | XP_011519754.1 |
NPTN-IT1 - NPTN intronic transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
REC114 - REC114 meiotic recombination protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |