Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACTGAAATCACTGCTCGTGCCAT[A/G]AACAGAAGGTAGGCTGGTGGATTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607511 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS17 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAMTS17 - ADAM metallopeptidase with thrombospondin type 1 motif 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139057.3 | 7065 | UTR 3 | NP_620688.2 | |||
XM_005254872.3 | 7065 | UTR 3 | XP_005254929.1 | |||
XM_011521312.2 | 7065 | UTR 3 | XP_011519614.1 | |||
XM_017021973.1 | 7065 | UTR 3 | XP_016877462.1 | |||
XM_017021974.1 | 7065 | Intron | XP_016877463.1 | |||
XM_017021975.1 | 7065 | UTR 3 | XP_016877464.1 | |||
XM_017021976.1 | 7065 | UTR 3 | XP_016877465.1 | |||
XM_017021977.1 | 7065 | Intron | XP_016877466.1 | |||
XM_017021978.1 | 7065 | UTR 3 | XP_016877467.1 | |||
XM_017021979.1 | 7065 | UTR 3 | XP_016877468.1 | |||
XM_017021980.1 | 7065 | UTR 3 | XP_016877469.1 | |||
XM_017021981.1 | 7065 | Intron | XP_016877470.1 | |||
XM_017021982.1 | 7065 | Intron | XP_016877471.1 | |||
XM_017021983.1 | 7065 | Intron | XP_016877472.1 | |||
XM_017021984.1 | 7065 | Intron | XP_016877473.1 |
LOC105371022 - uncharacterized LOC105371022 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |