Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGGAGGTGCCTCGAGAGGTGTGT[C/G]GGGGGCGCAGGTAGGTGCTGAAGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 612039 MIM: 608963 | ||||||||||||||||||||
Literature Links: |
LPCAT4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LPCAT4 - lysophosphatidylcholine acyltransferase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_153613.2 | 1558 | Missense Mutation | CCA,CGA | P,R 490 | NP_705841.2 | |
XM_006720454.2 | 1558 | Intron | XP_006720517.1 | |||
XM_011521422.1 | 1558 | Missense Mutation | CCA,CGA | P,R 322 | XP_011519724.1 |
NUTM1 - NUT midline carcinoma family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |