Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCCAGAAATTTCAGCTGCAGTTA[G/T]GAAGATTTTTGATGCTGCCAAAGTA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 614362 MIM: 601149 | ||||||||||||||||||||
Literature Links: |
ACSBG1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACSBG1 - acyl-CoA synthetase bubblegum family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IDH3A - isocitrate dehydrogenase 3 (NAD(+)) alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005530.2 | 194 | Missense Mutation | AGG,ATG | R,M 51 | NP_005521.1 | |
XM_005254336.2 | 194 | UTR 5 | XP_005254393.1 | |||
XM_017022118.1 | 194 | Missense Mutation | AGG,ATG | R,M 1 | XP_016877607.1 | |
XM_017022119.1 | 194 | UTR 5 | XP_016877608.1 | |||
XM_017022120.1 | 194 | UTR 5 | XP_016877609.1 |