Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCAACTTTTGGCTATACCCTACTG[G/T]CAGCCATAGTAGAGAGAGCTTCAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608440 | ||||||||||||||||||||
Literature Links: |
LACTB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LACTB - lactamase beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288585.1 | 1066 | Missense Mutation | TGG,TGT | W,C 330 | NP_001275514.1 | |
NM_032857.4 | 1066 | Missense Mutation | GCA,TCA | A,S 332 | NP_116246.2 | |
NM_171846.3 | 1066 | Missense Mutation | GCA,TCA | A,S 332 | NP_741982.1 | |
XM_011521180.1 | 1066 | Missense Mutation | GCA,TCA | A,S 332 | XP_011519482.1 |