Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATTTTAGCCTTGATCGAAAAAA[A/G]AATCCAGAACCTTGGGAAACTGTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608409 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C15orf48 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C15orf48 - chromosome 15 open reading frame 48 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032413.3 | 292 | Silent Mutation | AAA,AAG | K,K 46 | NP_115789.1 | |
NM_197955.2 | 292 | Silent Mutation | AAA,AAG | K,K 46 | NP_922946.1 |
MIR147B - microRNA 147b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |