Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCAAGAAACATCCAAGTGCCTC[A/G]ATGTCATGCCAAAGGCAGCAGCAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
LOC105370888 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105370888 - uncharacterized LOC105370888 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM202 - transmembrane protein 202 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080462.1 | 117 | Silent Mutation | TCA,TCG | S,S 39 | NP_001073931.1 | |
XM_011521497.1 | 117 | UTR 5 | XP_011519799.1 | |||
XM_011521499.1 | 117 | Intron | XP_011519801.1 |