Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTGCAGAGGTTAAAACACCTTTT[C/G]ATTTGGCCAAGGTGAGTGCTTATAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610696 MIM: 604501 | ||||||||||||||||||||
Literature Links: |
KIAA0101 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA0101 - KIAA0101 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TRIP4 - thyroid hormone receptor interactor 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321924.1 | 454 | UTR 5 | NP_001308853.1 | |||
NM_016213.4 | 454 | Missense Mutation | CAT,GAT | H,D 132 | NP_057297.2 |